Learn More
3M™ PELTOR™ Optime™ I Earmuffs
Description
- Low-profile design which minimizes obtrusiveness
- Large space inside cup helps reduce moisture and heat build-up
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
Specifications
Specifications
| Color | Hi-Viz |
| Certifications/Compliance | CE Approved, EN 352-3: 2002 |
| Model | H510F-459-GB |
| Noise Reduction Rating (NRR) | 28 dB |
| Product Type | Ear Muffs |
| Style | Over-the-head |
| Foldable | Yes |