Learn More
3M™ PELTOR™ CH-3 Listen Only Headset
PELTOR™ CH-3 Listen Only Headsets utilise the FLX2 cable system for a simple and reliable connection. They feature a speaker system that’s optimised for speech frequency in high noise environments and a modern cup design which provides high comfort and attenuation.
25330.00 NOK
Specifications
Color Blue
Product Type Communication Headset
Noise Level 25 dB
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17879066
View Documents Promotion Details
3M™
7100203297
Hard Hat
25330.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
17889066
View Documents Promotion Details
3M™
7100203639
25330.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 16-07-2024
Description
  • Protect your hearing without losing the ability to hear conversation with PELTOR™ CH-3 Listen Only Headsets.
  • These hear-only headsets feature a CH-3 speaker system that’s optimized for the speech frequency in high noise environments and meets the efficiency demand from today's digital devices.
  • The FLX2 cable system provides a simple and reliable connection and the modern cup design offers high comfort and attenuation.
  • All critical parts are rigorously tested and easy to change without any special tooling which contributes to low maintenance costs.
  • Comes with a headband, neckband or helmet attachment.
  • Available in bright yellow for the standard model or bright blue to indicate Explosive atmosphere (Ex) approval.
Specifications
Specifications
Blue
Communication Headset
25 dB