Learn More
3M™ PELTOR™ Alert FM-Radio Headset
Product Code. 15779987 Shop All 3M Products
Change view
Click to view available options
Product Type:
Fortified Safety Helmet, Radio Headset
Radio Headset
Model:
Fortified Safety Helmet
Headband
Weight (Metric):
400 g
430 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Model Weight (Metric) Unit Size
15779987 Radio Headset Headband 400 g Each
15739997 Fortified Safety Helmet, Radio Headset Fortified Safety Helmet 430 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15779987 Supplier 3M™ Supplier No. 7000108376
  • ONLINE EXCLUSIVE PRICE
    3820.00 NOK
    1804.23 NOK / Pack of 1
    Save 2015.77 NOK
    52% Off
Estimated Shipment: 15-01-2027
3M™ PELTOR™ Alert FM-Radio Headset with built in FM radio and a level dependent function to improve safety and increase motivation for wearers in noisy conditions.
  • Protect your hearing and listen to the radio - at the same time
  • Excellent FM-radio reception
  • Level dependent function lets the user hear surrounding noise and converse with colleagues
  • 3.5 mm stereo audio input plug to connect to external devices
  • Operating time approx. 70 hours
  • Flex system means you only need replace the cable if you update or change your radio manufacturer, not the expensive part, the headset
TRUSTED_SUSTAINABILITY
Specifications
Product Type Radio Headset
Model Headband
Noise Level 30 dB
Weight (Metric) 400 g
Certifications/Compliance CE Approved