Learn More
Peimisine, MedChemExpress
Product Code. 16451338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16451338
Brand: Medchem Express HYN0214
MedChemExpress Peimisine (Ebeiensine) is a muscarinic M receptor antagonist and angiotensin converting enzyme (ACE) inhibitor. Peimisine shows anti-tumor, anti-inflammatory, antihypertensive activities. Peimisine can induce apoptosis and be used in cough and asthma research.
Peimisine (Ebeiensine) is a muscarinic M receptor antagonist and angiotensin converting enzyme (ACE) inhibitor. Peimisine shows anti-tumor, anti-inflammatory, antihypertensive activities. Peimisine can induce apoptosis and be used in cough and asthma research.
Specifications
Chemical Name or Material Peimisine
CAS 19773-24-1
Color White
Purity Grade Notes Research
Molecular Formula C27H41NO3
Quantity 5 mg
Synonym Ebeiensine
Solubility Information DMSO : 12.5 mg/mL (29.23 mM; ultrasonic and heat to 60°C)
SMILES O=C1[C@](C[C@@H](O)CC2)([H])[C@@]2(C)[C@@]3([H])CC4=C(C)[C@@]5(O[C@@]6([H])[C@@]([C@H]5C)([H])NC[C@@H](C)C6)CC[C@@]4([H])[C@]3([H])C1
Molecular Weight (g/mol) 427.62
Formula Weight 427.62
Percent Purity 98.47%
Grade Research
Physical Form Powder
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.