Learn More
Hanna Instruments™ Gel Filled PEI Body Combination pH Electrodes
missing translation for 'orderingAttributeHoverText'
Length (Metric):
120 mm
missing translation for 'unitSize'
Each
Description
- Is a PEI body, double junction pH electrode with a BNC connector.
- Electrode features a single ceramic junction in the outer reference cell
- Spherical pH sensing portion is made with general purpose glass and is protected by an extension of the durable PEI body
- Design consideration is ideal for field applications as well as general purpose use
- Uses general purpose (GP) glass, spherical bulb, polyetherimide (PEI) body, single ceramic frit, and double junction
Specifications
Specifications
| Connector Types | BNC |
| Diameter (Metric) | 12 mm |
| Length (Metric) | 120 mm |
| pH Range | 0 to 13 |
| Temperature (Metric) Range | 0°C to 80°C |