Description
Get excellent chemical compatibility and very low carryover to improve liquid chromatography (LC) performance using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing. Made of precision-bore fused silica tubing coated with a 1/16 in. O.D. PEEK covering, it is capable of withstanding high pressures. PEEKsil tubing is ideal for capillary HPLC and LC-mass spectrometry (MS). The very smooth internal surface ensures excellent flow characteristics and very low carryover. It is compatible with most organic solvents and is usable in most standard chromatography systems.
- Tubing is stiff; not recommended for uses requiring that tubing bends
- Use precut lengths only; tube cutting after receipt may damage tubing
Specifications
Specifications
| For Use With | HPLC, LC/MS |
| Type | Capillary Tubing |
| Description | 0.007 in. ID; 10 cm length |
| For Use With (Application) | HPLC, LC/MS |
| Diameter (English) Inner | 0.007 in. |
| Diameter (English) Outer | 1/16 in. |
| Diameter (Metric) Inner | 0.18 mm |
| Diameter (Metric) Outer | 1.588 mm |
| Overall Length (English) | 3.937 in. |
| Overall Length (Metric) | 10 cm |
| Show More |