Learn More
Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections
Shop All Thermo Scientific ProductsDescription
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
- Absolute zero or low dead volume formats
- Biocompatible
Specifications
Specifications
| Type | Cross |
| Diameter (English) Inner | 0.02 in. |
| Description | PEEK Cross; <0.72 uL Swept Volume |
| Material | PEEK |
| For Use With (Equipment) | 1/16 in. OD Tubing |
| Length (Metric) | 27.69 mm |