Learn More
Cytiva PEEK Union
Product Code. 12646185 Shop All Cytiva Products
Change view
Click to view available options
Connections:
1/16 Male
1/16 in. Female to 1/16 in. Female
5/16 in. Female to 1/16 in. Male
M6 female to 1/16 in. Male
Unit Size:
Pack of 5
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Unit Size
12646185 1/16 in. Female to 1/16 in. Female Pack of 5
12534317 5/16 in. Female to 1/16 in. Male Pack of 8
11340072 M6 female to 1/16 in. Male Pack of 5
12465610 1/16 Male Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12646185 Supplier Cytiva Supplier No. 11000339
Estimated Shipment: 16-11-2026
Connect piping of varying sizes. Cytiva Life Sciences™ PEEK Unions are an important accessory for various scientific applications.
  • Packsize 5
  • When ordered separately the connectors are black.
  • Made of polyetheretherketone (PEEK)
TRUSTED_SUSTAINABILITY
Specifications
Type Union
Connections 1/16 in. Female to 1/16 in. Female
Material PEEK