Learn More
Saint Gobain Life Sciences Versilon™ PEEK Tubing
Product Code. p-8004371
Click to view available options
Diameter (Metric) Inner:
0.51 mm
0.76 mm
Color:
Green
Orange
Unit Size:
1.5m
Product Code. 11716319
Brand: Saint Gobain Life Sciences 0200630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Versilon™ PEEK Tubing is flexible, capillary tubing designed specifically for the demands of high-pressure chromatography (HPLC).
Versilon™ PEEK flexible, capillary tubing from Saint-Gobain is designed specifically for the demands of high-pressure chromatography (HPLC). More corrosion resistant than stainless steel, this tubing is strong enough to take the pressure of 5000 psi. Versilon™ PEEK tubing's heat and chemical resistance properties are similar to PTFE, but it is structurally strong and abrasion resistant. It does not leach, and it is unaffected by continuous exposure to steam.
  • High-pressure demands of chromatography – up to 5000 psi
  • Corrosion-resistant
  • Heat resistance
  • Chemical resistance
  • Abrasion-resistant
  • Does not leach
  • Unaffected by continuous exposure to steam
Typical Applications
  • Semiconductor industry
  • Liquid chromatography
  • Pump and valve components
  • Seal rings
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Tubing
Autoclavable Autoclavable
Color Green
For Use With (Application) Chromatography
Diameter (English) Inner 0.03 in.
Diameter (English) Outer 0.06 in.
Diameter (Metric) Inner 0.76 mm
Diameter (Metric) Outer 1.6 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.52 m
Material PEEK
Maximum Pressure 3000 psi
Max. Operating Temperature (English) 212°F
Min. Operating Temperature (Metric) 100°C
Max. Operating Temperature (Metric) 100°C