Learn More
Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16 in. O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
Specifications
Specifications
| For Use With | Fused Silica Capillary Tubing |
| Type | Capillary Tubing Sleeve |
| Description | 0.010 in. ID; Blμe |
| Diameter (English) Inner | 0.010 in. |
| Diameter (Metric) Inner | 0.25 mm |
| Material | PEEK |