Learn More
Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing
Product Code. 10220585 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
0.008 in. ID; Yellow
0.010 in. ID; Blμe
0.012 in. ID; Natural
0.015 in. ID; Orange
0.021 in. ID; Natural
0.030 in.ID; Natural
Unit Size:
Each
Product Code. Description unitSize
10220585 0.012 in. ID; Natural Each
10372035 0.008 in. ID; Yellow Each
10656824 0.010 in. ID; Blμe Each
10771654 0.015 in. ID; Orange Each
10611416 0.021 in. ID; Natural Each
10147574 0.030 in.ID; Natural Each
6 options
Product Code. 10220585
Brand: Thermo Scientific™ F229
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make capillary tubing connections with Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing.
Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16 in. O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Fused Silica Capillary Tubing
Type Capillary Tubing Sleeve
Description 0.012 in. ID; Natural
Diameter (English) Inner 0.012 in.
Diameter (Metric) Inner 0.305 mm
Material PEEK