Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Product Code. 10618915 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Fingertight Fitting
One-Piece PEEK Fingertight Fitting
One-Piece PEEK Long Fingertight Fitting
PEEK Wing Nut w/PEEK Ferrule
Replacement Ferrule
Short Head Port Plug, Black
Short Head Port Plug, Red
Description:
1/16 in.
1/16 in., 0.37 in. Head
1/16 in., 10-32 Coned, Delrin, Black
1/16 in., 10-32 Coned, Delrin, Red
PEEK
PEEK, 1/32 in., 0.25 in. Head
Unit Size:
Each
Pack of 10
Product Code. Type Description unitSize
10618915 Replacement Ferrule PEEK Pack of 10
10706163 One-Piece PEEK Fingertight Fitting 1/16 in., 0.37 in. Head Pack of 10
10754994 One-Piece PEEK Long Fingertight Fitting 1/16 in., 0.37 in. Head Pack of 10
11503980 PEEK Wing Nut w/PEEK Ferrule 1/16 in. Each
10504465 Fingertight Fitting PEEK, 1/32 in., 0.25 in. Head Pack of 10
15461073 Short Head Port Plug, Black 1/16 in., 10-32 Coned, Delrin, Black Each
15319849 Short Head Port Plug, Red 1/16 in., 10-32 Coned, Delrin, Red Each
7 options
Product Code. 10618915
Brand: Thermo Scientific™ F142X
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Ferrule
Description PEEK
For Use With (Equipment) LC Tubing
Material PEEK
Quantity 10 Pack