Learn More
Thermo Scientific™ PEEK Capillary Tubing
Product Code. 10069382 Shop All Thermo Scientific Products
Change view
Click to view available options
Overall Length (Metric):
5 cm
10 cm
20 cm
30 cm
50 cm
100 cm
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
Color:
Blue
Natural
Orange
Red
Yellow
Unit Size:
Each
Product Code. Overall Length (Metric) Diameter (English) Inner Color unitSize
10069382 30 cm 0.005 in. Red Each
10259914 10 cm 0.003 in. Natural Each
10280195 100 cm 0.003 in. Natural Each
10302185 20 cm 0.003 in. Natural Each
10794267 30 cm 0.003 in. Natural Each
10006604 5 cm 0.003 in. Natural Each
10678515 50 cm 0.003 in. Natural Each
10552265 10 cm 0.005 in. Red Each
10609993 20 cm 0.005 in. Red Each
10573225 5 cm 0.005 in. Red Each
10766524 50 cm 0.005 in. Red Each
10498084 100 cm 0.007 in. Yellow Each
10381485 20 cm 0.007 in. Yellow Each
10594665 30 cm 0.007 in. Yellow Each
10439244 50 cm 0.007 in. Yellow Each
10249414 10 cm 0.010 in. Blue Each
10504255 100 cm 0.010 in. Blue Each
10574855 30 cm 0.010 in. Blue Each
10752917 5 cm 0.010 in. Blue Each
10076304 50 cm 0.010 in. Blue Each
10581305 100 cm 0.020 in. Orange Each
10161033 20 cm 0.020 in. Orange Each
10464793 30 cm 0.020 in. Orange Each
10180553 50 cm 0.020 in. Orange Each
24 options
Product Code. 10069382
Brand: Thermo Scientific™ 3700530
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors.
Use Thermo Scientific™ Rheodyne™ Sample Loops for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With Rheodyne HPLC Sample Injectors
Type Capillary Tubing
Color Red
Diameter (English) Inner 0.005 in.
Diameter (Metric) Inner 0.127 mm
Overall Length (English) 11.811 in.
Overall Length (Metric) 30 cm
Material PEEK