Learn More
Hughes Pedestal Mounted Open Bowl Eye/Face Wash
Product Code. 17732338
Change view
Click to view available options
Mounting:
Pedestal
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
17732338 Pedestal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17732338 Supplier Hughes Supplier No. HEWP0085SS
  • ONLINE EXCLUSIVE PRICE
    7061.25 NOK
    4707.93 NOK / Pack of 1
    Save 2353.32 NOK
    33% Off
Estimated Shipment: 19-10-2026
Open stainless steel bowl, pedestal-mounted emergency eye/face wash featuring twin spray nozzles protected by dust covers which lift off upon activation via the push plate.
  • Open stainless steel bowl, pedestal-mounted emergency eye/face wash featuring twin spray nozzles protected by dust covers which lift off upon activation via the push plate
  • The flow regulator valve guarantees an optimum water flow
  • The eye wash diffusers are fitted with fine mesh strainers and individual flow controllers designed to give a gentle flow of aerated water at 12 liters per minute as per the ANSI standard
  • This unit is intended for indoor use but can be located outdoors where there is no risk of overheating or freezing
Specifications
Height (Metric) 35 cm
Material Stainless Steel
Mounting Pedestal
Product Type Emergency Shower
Width (Metric) 38 cm