Learn More
Hughes Pedestal Mounted Closed Bowl Eye/Face Wash
Product Code. 17722338
Change view
Click to view available options
Mounting:
Pedestal
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
17722338 Pedestal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17722338 Supplier Hughes Supplier No. HEWP0045GS
  • ONLINE EXCLUSIVE PRICE
    13815.98 NOK
    10391.36 NOK / Pack of 1
    Save 3424.62 NOK
    24% Off
Estimated Shipment: 14-12-2026
Pedestal mounted emergency eye/face wash.
  • Pedestal mounted emergency eye/face wash
  • This ANSI compliant model has an integral lid to protect the bowl and diffusers against dust and debris
  • Designed to be quick and easy to operate with high flow rate aerated diffusers for improved washing
Specifications
Depth (Metric) 46 cm
Material Stainless Steel
Mounting Pedestal
Product Type Emergency Shower
Width (Metric) 32 cm