Learn More
Gilac™ Pedal Container with Lid
Click to view available options
Closure Color:
Blue
Green
White
Unit Size:
Each
Description
- Equipped with two antimark wheels (diameter 100 mm)
- Easy to move around
- Hands-free operation for sanitary disposal
- Easy to clean
- Coloured lids for selective waste sorting
Specifications
Specifications
| Capacity (Metric) | 100 L |
| Closure Color | White |
| Capacity (English) | 26.4 gal. |
| Color | White |
| Dimensions (L x W x H) | 515 x 477 x 925 mm |
| Material | Polypropylene |