missing translation for 'onlineSavingsMsg'
Learn More

PECI Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20383-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849914

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PECI is an auxiliary enzyme that catalyzes an isomerization step required for the beta-oxidation of unsaturated fatty acids.
Specifications

Specifications

PECI
Polyclonal
Unconjugated
ECI2
ACBD2, D3,D2 enoyl CoA isomerase, DBI related protein 1, Dodecenoyl CoA isomerase, DRS 1, DRS1, HCA88, KIAA0536, PECI
Rabbit
Antigen Affinity Chromatography
RUO
10455, 23986
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75521, Q9WUR2
ECI2
PECI Fusion Protein Ag14211
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers