Learn More
Tork PeakServe™ Mini Continuous Paper Hand Towel Dispenser H5
Product Code. 30243822 Shop All Tork Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243822 Black Each
30243800 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243822 Supplier Tork Supplier No. 552558
  • ONLINE EXCLUSIVE PRICE
    1964.93 NOK
    1755.07 NOK / Pack of 1
    Save 209.86 NOK
    10% Off
Estimated Shipment: 21-12-2026
The latest innovation designed for high-traffic washrooms where staff need to maximise cleaning time and maintain a good flow of people.
  • Say goodbye to queues in busy washrooms with this paper towel dispenser that delivers a towel in just three seconds.
  • Prevent hand towels running out with 250% more capacity than Tork Xpress Multifold Paper Towel dispensers.
  • Cut refilling time in half and spend more time on other tasks with a hand towel dispenser you can refill at any point.
  • Avoid towel jams or towel tabbing with our patented Continuous system for paper towels.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Towel Dispenser
Color Black
Alternate Part Number 7322541136585
Holds Hand Towels
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid
Model Elevation