Learn More
asecos™ PE IBC Station with Filling Area and Galvanised Grid
Product Code. 17826386 Shop All asecos Products
Change view
Click to view available options
Width (Metric):
1280.00 mm
2460.00 mm
3640.00 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
17826386 1280.00 mm Each
17876426 2460.00 mm Each
17826346 3640.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17826386 Supplier asecos™ Supplier No. 24156
This item requires a price update prior to being ordered.
Request a Price for this item here.
The PE IBC Station is approved for the safe storage of water pollutants.
  • Highly resistant against oil, acid, alkali, etc.
  • Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Color Blue
Capacity (English) 264.2 gal.
Capacity (Metric) 1000 L
Material Polyethylene
Type IBC Spill Pallet
Length (English) 50.39 in.
Length (Metric) 1280.00 mm
Width (English) 50.39 in.
Width (Metric) 1280.00 mm
Height (English) 33.86 in.
Height (Metric) 860.00 mm
Depth (English) 75.20 in.
Depth (Metric) 1910.00 mm
Weight (English) 253.53 lb.
Weight (Metric) 115 kg