missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDK3 Polyclonal antibody specifically detects PDK3 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | PDK3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11.2, Pyruvate dehydrogenase kinase isoform 3, pyruvate dehydrogenase kinase, isoenzyme 3, pyruvate dehydrogenase kinase, isozyme 3, pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 137-406 of human PDK3 (NP_005382.1).,, Sequence:, FGFDPFISTNIQYFLDRFYTNRISFRMLINQHTLLFGGDTNPVHPKHIGSIDPTCNVADVVKDAYETAKMLCEQYYLVAPELEVEEFNAKAPDKPIQVVYVPSHLFHMLFELFKNSMRATVELYEDRKEGYPAVKTLVTLGKEDLSIKISDLGGGVPLRKIDRLFNYMYSTAPRPSLEPTRAAPLAGFGYGLPISRLYARYFQGDLKLYSMEGVGTDAVIYLKALSSESFERLPVFNKSAWRHYKTTPEADDWSNPSSEPRDASKYKAKQ |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?