missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRA Monoclonal Antibody (16A1), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110099
Additional Details : Weight : 0.01000kg
Description
This gene encodes a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes. [provided by RefSeq, Jul 2008]Specifications
PDGFRA | |
Monoclonal | |
PE | |
P16234 | |
PDGFRA | |
CD140a-transfected NIH 3T3 cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
16A1 | |
PDGFRA | |
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a | |
Mouse | |
Purified | |
RUO | |
5156 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |