Learn More
PDGF R beta Antibody (APB5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-43349-0.05MG
 View more versions of this product
Product Code. 18296893
  • 3350.00 NOK / 0.05mg
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PDGF R beta
Monoclonal
0.5 mg/mL
Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P05622
PDGFRB
The immunogen for this antibody was PDGF receptor beta.
0.05 mg
Protein Kinase
5159
Store at 4C. Do not freeze.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
APB5
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Rat
Affinity Purified
RUO
Primary
Mouse
IgG2a κ