Learn More
PDE7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17771-1-AP-150UL
This item is not returnable.
View return policy
Description
The 3′, 5′-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3′, 5′-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5′-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a cAMP-specific phosphodiesterase, a member of the cyclic nucleotide phosphodiesterase family.Specifications
| PDE7B | |
| Polyclonal | |
| Unconjugated | |
| PDE7B | |
| PDE7B, phosphodiesterase 7B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140929, 27115, 29863 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NP56, Q9QXQ1 | |
| PDE7B | |
| PDE7B Fusion Protein Ag12184 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |