missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE7A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26277-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE7 subfamily. This PDE hydrolyzes the second messenger, cAMP, which is a regulator and mediator of a number of cellular responses to extracellular signals. Thus, by regulating the cellular concentration of cAMP, this protein plays a key role in many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
PDE7A | |
Polyclonal | |
Unconjugated | |
PDE7A | |
HCP1, PDE7, PDE7A, phosphodiesterase 7A, TM22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5150 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13946 | |
PDE7A | |
PDE7A Fusion Protein Ag24334 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |