missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16226-1-AP-150UL
This item is not returnable.
View return policy
Description
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.Specifications
| PDE4A | |
| Polyclonal | |
| Unconjugated | |
| PDE4A | |
| DPDE2, PDE4, PDE46, PDE4A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18577, 25638, 5141 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O89084, P27815, P54748 | |
| PDE4A | |
| PDE4A Fusion Protein Ag8018 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction