Learn More
PDE10A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18078-1-AP-20UL
This item is not returnable.
View return policy
Description
The PDE10A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PDE10A. This antibody recognizes human, mouse, rat antigen. The PDE10A antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
| PDE10A | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 10846 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunofluorescence | |
| 240 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |