missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14997-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.Specifications
PDCL3 | |
Polyclonal | |
Unconjugated | |
PDCL3 | |
HTPHLP, PDCL3, PHLP2A, PHLP3, phosducin like 3, Phosducin like protein 3, PhPL3, VIAF, VIAF 1, VIAF1, Viral IAP associated factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68833, 79031 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BVF2, Q9H2J4 | |
PDCL3 | |
PDCL3 Fusion Protein Ag6983 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |