missing translation for 'onlineSavingsMsg'
Learn More

PDCD4 Mouse anti-Human, Clone: 1F1A11, Proteintech

Mouse Monoclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen PDCD4
Clone 1F1A11
Concentration 0.74 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887665
View Documents
Proteintech
66100-1-IG-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16877665
View Documents
Proteintech
66100-1-IG-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.
Specifications

Specifications

PDCD4
0.74 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a
PDCD4
IgG1
Protein G
Antibody
1F1A11
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
Q53EL6
27250
PDCD4 Fusion Protein Ag19239
Primary
-20°C
PDCD4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.