missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15081-1-AP-150UL
This item is not returnable.
View return policy
Description
PDGFA-associated protein 1 (PDAP1) is a phosphoprotein that may upregulate the PDGFA-stimulated growth of fibroblasts and also downregulate the mitogenicity of PDGFB. PDGFA in rodents has been shown to bind PDGFA with a low affinity.Specifications
| PDAP1 | |
| Polyclonal | |
| Unconjugated | |
| PDAP1 | |
| HASPP28, PAP, PAP1, PDAP1, PDGF associated protein, PDGFA associated protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11333 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13442 | |
| PDAP1 | |
| PDAP1 Fusion Protein Ag7155 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction