missing translation for 'onlineSavingsMsg'
Learn More

PD 151746, MedChemExpress

Product Code. p-200096321
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477696 100 mg 100mg
16463686 25 mg 25mg
16460348 5 mg 5mg
16427888 10 mg 10mg
16403837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477696 Supplier Medchem Express Supplier No. HY19749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD151746 is a calpain inhibitor, shows a 20-fold selectivity for u-calpain (Ki = 0.26 ± 0.03 μM) over m-calpain (Ki = 5.33 ± 0.77 μM).

PD151746 is a calpain inhibitor, shows a 20-fold selectivity for u-calpain (Ki = 0.26 ± 0.03 μM) over m-calpain (Ki = 5.33 ± 0.77 μM).

Specifications

Chemical Name or Material PD 151746
CAS 181765-30-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8FNO2S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (210.75 mM; Need ultrasonic)
SMILES O=C(O)/C(S)=C/C1=CNC2=C1C=C(F)C=C2.[(Z)]
Molecular Weight (g/mol) 237.25
Formula Weight 237.25
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.