Learn More
PD 117519, MedChemExpress
Product Code. 16484216
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484216 100 mg 100mg
16475497 5 mg 5mg
16469456 50 mg 50mg
16462758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16484216 Supplier Medchem Express Supplier No. HY100032
MedChemExpress PD 117519 (CI947) is an A2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models.
PD 117519 (CI947) is an A2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models.
Specifications
Chemical Name or Material PD 117519
CAS 96392-15-3
Color White
Purity Grade Notes Research
Molecular Formula C19H21N5O4
Quantity 100 mg
Synonym CI947
Solubility Information DMSO : ≥ 100 mg/mL (260.82 mM)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N[C@@H]4CCC5=C4C=CC=C5)O1)O)O
Molecular Weight (g/mol) 383.4
Formula Weight 383.4
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation