Learn More
PD-1 Antibody (1015829), HRP, R&D Systems™
Product Code. 18605933 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18605933 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18605933 Supplier Novus Biologicals Supplier No. FAB10868H
  • ONLINE EXCLUSIVE PRICE
    5487.12 NOK
    4710.30 NOK / 0.1mL
    Save 776.82 NOK
    14% Off
 Restricted Item
Estimated Shipment: 26-11-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Applications Blockade of Receptor-ligand Interaction, Flow Cytometry
Product Type Antibody
Product Line Novus