missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PCSK2 (Human) Recombinant Protein
Human PCSK2 full-length ORF ( AAH40546, 1 a.a. - 619 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005126-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVFASAERPVFTNHFLVELHKGGEDKARQVAAEHGFGVRKLPFAEGLYHFYHNGLAKAKRRRSLHHKQQLERDPRVKMALQQEGFDRKKRGYRDINEIDINMNDPLFTKQWYLINTGQADGTPGLDLNVAEAWELGYTGKGVTIGIMDDGIDYLHPDLASNYNAEASYDFSSNDPYPYPRYTDDWFNSHGTRCAGEVSAAANNNICGVGVAYNSKVAGIRMLDQQFMTDIIEASSISHMPQLIDIYSASWGPTDNGKTVDGPRELTLQAMADGVNKGRGGKGSIYVWASGDGGSYDDCNCDGYASSMWTISINSAINDGRTALYDESCSSTLASTFSNGRKRNPEAGVATTDLYGNCTLRHSGTSAAAPEAAGVFALALEANLGLTWRDMQHLTVLTSKRNQLHDEVHQWRRNGVGLEFNHLFGYGVLDAGAMVKMAKDWKTVPERFHCVGGSVQDPEKIPSTGELVLTLTTDACEGKENFVRYLEHVQAVITVNATRRGDLNINMTSPMGTKSILLSRRPRDDDSKVGFDKWPFMTTHTWGEDARGTWTLELGFVGSAPQKGVLKEWTLMLHGTQSAPYIDQVVRDYQSKLAMSKKEELEEELDEAVERSLKSILNKN
Specifications
AAH40546 | |
proprotein convertase subtilisin/kexin type 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCSK2 | |
GST |
5126 | |
Wheat germ expression system | |
25 μg | |
NEC2, PC2, SPC2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |