Learn More
Microspec™ PCR Tubes
Description
- PCR tubes
- Natural
- Pack of 1000
Specifications
Specifications
| Closure Color | Clear |
| Color | Clear |
| Volume (Metric) | 0.5 mL |
| Storage Requirements | Room Temperature |
| Quantity | 1000 |
| Closure Material | Polypropylene |
| Shape | Conical |
| Sterility | Sterile |
| Material | Polypropylene |
| Dnase Free | No |
| Show More |