Learn More
BRAND™ PCR Tube Strips
Product Code. p-8011224 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
Green
Rose
White
Volume (Metric):
0.15 mL
0.2 mL
Unit Size:
Pack of 120
Pack of 125
Pack of 250
Product Code. Color Volume (Metric) unitSize
11688419 White 0.15 mL Pack of 120
11678419 Clear 0.2 mL Pack of 250
10789064 Green 0.2 mL Pack of 125
10478092 Rose 0.2 mL Pack of 125
11618439 White 0.2 mL Pack of 120
5 options
Product Code. 11688419
Brand: BRAND™ 781317
Extra-thin and even wall thicknesses allow for optimal heat transfer and short cycle times. Smooth shaft surfaces minimize enzyme and nucleic acid binding to the surface ensuring the reaction can run in full.
  • Polypropylene 8 and 12 tube strips
  • A variety of cap styles is available
  • Compatible with leading thermal cyclers with heated lids
  • Provide tight seal in all PCR* cyclers
  • Protect samples from cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Transparent
Color White
Volume (Metric) 0.15 mL
Quantity 120 Pack
Material Polypropylene
Closure Type Flat Cap (Attached)
Product Type PCR Tube Strip