Learn More
MP Biomedicals™ PCR Kit Core Kit 25
Product Code. 11487030 Shop All MP Biomedicals Products
Product Code. 11487030
Brand: MP Biomedicals™ 11EPTQK255C
Taq and dNTPS in one kit.
  • All the reagents for PCR buffers included
  • NTPs at 25mM each
  • Product shipped on dry ice 5 x 250 units
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 x 250U