Includes
96-well PCR plates, PCR-sealing film
Premium PCR/qPCR transparent plates
- 96-well
- Skirted, non-skirted, or semi-skirted
- Low, standard or elevated rim profile
- Ultra-thin walled for quick and constant thermal transfer
- Free of DNase, DNA, RNase and endotoxins
- No warping, no wiggling, suitable for automation
- Highly transparent for qPCR
- Press to seal
- Convenient end tabs
- No annoying adhesive residues
Specifications
Specifications
| Description | Elevated rim low profile plates |
| Quantity | 50/Pk. |
| No. of Wells | 96 |
| Format | 96-well |
| Product Type | PCR Combo Pack |