Learn More
PCNP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11180-2-AP-150UL
This item is not returnable.
View return policy
Description
PCNP is a protein coding gene, and may be involved in cell cycle regulation.Specifications
| PCNP | |
| Polyclonal | |
| Unconjugated | |
| PCNP | |
| PCNP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288165, 57092, 76302 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6P8I4, Q7TP40, Q8WW12 | |
| Pcnp | |
| PCNP Fusion Protein Ag1663 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |