Learn More
PCIF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16082-1-AP-150UL
 View more versions of this product
Product Code. 16845564
  • 4836.10 NOK / 150µL
Description
PCIF1 may play a role in transcription elongation or in coupling transcription to pre-mRNA processing through its association with the phosphorylated C-terminal domain (CTD) of RNAPII largest subunit.
Specifications
Specifications
PCIF1
Polyclonal
Unconjugated
PCIF1
C20orf67, PCIF1
Rabbit
Antigen Affinity Chromatography
RUO
228866, 63935
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59114, Q9H4Z3
PCIF1
PCIF1 Fusion Protein Ag9080
150 μL
Primary
Human, Mouse