Learn More
PCID2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26638-1-AP-150UL
This item is not returnable.
View return policy
Description
The PCID2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PCID2. This antibody recognizes human antigen. The PCID2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| PCID2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| 150 μL | |
| 55795 | |
| IgG |