missing translation for 'onlineSavingsMsg'
Learn More

PCI-27483, MedChemExpress

Product Code. 16427048
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16427048

Brand: Medchem Express HY16382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PCI-27483 is a FVIIa/tissue factor inhibitor, with antitumour effects.

PCI-27483 is a FVIIa/tissue factor inhibitor, with antitumour effects.

Specifications

Chemical Name or Material PCI-27483
CAS 871266-63-6
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C26H24N6O9S
Quantity 10 mg
Solubility Information DMSO : 62.5 mg/mL (104.77 mM; Need ultrasonic)
SMILES O=C(O)C[C@@H](C(O)=O)NC(CC1=CC(C2=CC(S(=O)(N)=O)=CC=C2O)=C(O)C(C3=NC4=CC=C(C(N)=N)C=C4N3)=C1)=O
Molecular Weight (g/mol) 596.57
Formula Weight 596.57
Percent Purity 95.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.