Learn More
PCDHB12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16822484 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16822484 150 μL 150µL
16832484 20 μL 20µL
2 options
Product Code. 16822484
Brand: Proteintech 140201AP150UL
Rabbit Polyclonal Antibody
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.
Specifications
Antigen PCDHB12
Applications Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCDHB12
Gene Accession No. Q9Y5F1
Gene Alias PCDH beta 12, PCDH BETA12, PCDHB12, protocadherin beta 12
Gene Symbols PCDHB12
Host Species Rabbit
Immunogen PCDHB12 Fusion Protein Ag5135
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56124, 93883
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG