missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25090-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the protocadherin family, and cadherin superfamily, of transmembrane proteins containing cadherin domains. These proteins mediate cell adhesion in neural tissues in the presence of calcium. The encoded protein may be involved in signaling at neuronal synaptic junctions. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes multiple cadherin domains and a transmembrane region. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
PCDH9 | |
Polyclonal | |
Unconjugated | |
PCDH9 | |
PCDH9, protocadherin 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211712, 306091, 5101 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HC56 | |
Pcdh9 | |
PCDH9 Fusion Protein Ag18696 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |