missing translation for 'onlineSavingsMsg'
Learn More

PCDH11Y Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20069-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889544

  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody can recognize PCDH11X and PCDH11Y.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
Specifications

Specifications

PCDH11Y
Polyclonal
Unconjugated
PCDH11Y
PCDH PC, PCDH Y, PCDH11, PCDH11X, PCDH11Y, PCDH22, PCDHY, Protocadherin 11, protocadherin 11 Y linked, Protocadherin 22, Protocadherin PC, Protocadherin prostate cancer
Rabbit
Antigen Affinity Chromatography
RUO
27328, 83259
-20°C
Liquid
Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BZA7, Q9BZA8
PCDH11X, PCDH11Y
Peptide
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers