Learn More
PCB Mixture for Food & Human Tissue Analysis 297 10 μg/mL in Isooctane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 32598-13-3, 38380-01-7, 37680-73-2, 32598-14-4, 38380-03-9, 74472-37-0 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Isooctane |