Learn More
Lenz Laborglasinstrumente™ PBT Red Screw Caps with Aperture
Brand:  Lenz Laborglasinstrumente™ 01321032
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10020261
  • 250.16 NOK / Pack of 10
Estimated Shipment: 09-09-2024
Specifications
Screw thread tubes
PBT
10Pack
Cap
Screw Cap
GL 32
DIN thread (GL) and aperture
180°C
PBT
Red
PBT
20 mm