missing translation for 'onlineSavingsMsg'
Learn More

PBRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16850334 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16850334 20 μL 20µL
16840334 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16850334

Brand: Proteintech 125631AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This locus encodes a subunit of ATP-dependent chromatin-remodeling complexes. The encoded protein has been identified as in integral component of complexes necessary for ligand-dependent transcriptional activation by nuclear hormone receptors. Mutations at this locus have been associated with primary clear cell renal cell carcinoma.

Specifications

Antigen PBRM1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PBRM1
Gene Accession No. Q86U86, Q8BSQ9
Gene Alias BAF180, BRG1 associated factor 180, hPB1, PB1, PBRM1, polybromo 1, Polybromo 1D, Protein polybromo 1
Gene Symbols PBRM1
Host Species Rabbit
Immunogen PBRM1 Fusion Protein Ag3148
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306254, 55193, 66923
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.