Learn More
Pazufloxacin Mesylate, TRC
Specifications
Specifications
| Chemical Name or Material | Pazufloxacin Mesylate |
| CAS | 163680-77-1 |
| Molecular Formula | C16 H15 F N2 O4 . C H4 O3 S |
| Quantity | 100 mg |
| Synonym | 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(3S)-','methanesulfonate (1:1), 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(3S)-','monomethanesulfonate (9CI), 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(S)-','monomethanesulfonate, Pasil, Pazucross, Pazufloxacin mesilate, Pazufloxacin mesylate, Pazufloxacin methanesulfonate, T 3762 |
| InChI Formula | InChI=1S/C16H15FN2O4.CH4O3S/c1-7-6-23-14-11(16(18)2-3-16)10(17)4-8-12(14)19(7)5-9(13(8)20)15(21)22;1-5(2,3)4/h4-5,7H,2-3,6,18H2,1H3,(H,21,22);1H3,(H,2,3,4)/t7-;/m0./s1 |
| SMILES | C[C@H]1COc2c3N1C=C(C(=O)O)C(=O)c3cc(F)c2C4(N)CC4.CS(=O)(=O)O |
| Molecular Weight (g/mol) | 414.41 |
| Formula Weight | 414.0897 |