Learn More
Pazopanib Hydrochloride, TRC
Product Code. 30040860
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040860 250 mg 250mg
30041200 100 mg 100mg
30040747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30040860 Supplier Toronto Research Chemicals Supplier No. TRCP210926250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 635702-64-6
Molecular Formula C21 H23 N7 O2 S . Cl H
Molecular Weight (g/mol) 473.98
Synonym Benzenesulfonamide, 5-[[4-[(2,3-dimethyl-2H-indazol-6-yl)methylamino]-2-pyrimidinyl]amino]-2-methyl-, hydrochloride (1:1), Benzenesulfonamide, 5-[[4-[(2,3-dimethyl-2H-indazol-6-yl)methylamino]-2-pyrimidinyl]amino]-2-methyl-, monohydrochloride (9CI), 5-[[4-[(2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl]amino]-2-methylbenzenesulfonamide hydrochloride, GW 786034B, Pazopanib hydrochloride
IUPAC Name 5-[[4-[(2,3-dimethylindazol-6-yl)-methylamino]pyrimidin-2-yl]amino]-2-methylbenzenesulfonamide;hydrochloride
SMILES Cl.CN(c1ccc2c(C)n(C)nc2c1)c3ccnc(Nc4ccc(C)c(c4)S(=O)(=O)N)n3
Spezifikation
Quantity 250 mg
InChI Formula InChI=1S/C21H23N7O2S.ClH/c1-13-5-6-15(11-19(13)31(22,29)30)24-21-23-10-9-20(25-21)27(3)16-7-8-17-14(2)28(4)26-18(17)12-16;/h5-12H,1-4H3,(H2,22,29,30)(H,23,24,25);1H
Formula Weight 473.1401
Percent Purity >95%
Chemical Name or Material Pazopanib Hydrochloride
Grade HPLC