Learn More
Emma Safety Footwear Patrick XD Safety Shoes
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Specifications
| Design | Patrick XD |
| Material | Leather |
| Size | 45 |